Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077129.2 | NP_001070597.1 | 5 - 6 | 642 | 57 | |
| GenBank mRNA | BC112571.1 | - | 5 - 6 | 643 | 57 | |
More Information
Set Membership: |
|

OSGIN1
Genomic Map