Assay Details
| Gene Symbol: TST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177489.3 | NP_803455.1 | 1 - 2 | 761 | 86 | |
| GenBank mRNA | BC112580.1 | - | 1 - 2 | 762 | 86 | |
| BT020995.1 | - | 1 - 2 | 729 | 86 | ||
| M58561.1 | - | 1 - 2 | 682 | 86 | ||
More Information
Set Membership: |
|

TST
Genomic Map