Assay Details
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035051.3 | NP_001030223.2 | 5 - 6 | 621 | 84 | |
| GenBank mRNA | BC114072.1 | - | 5 - 6 | 622 | 84 | |
| BT021870.2 | - | 5 - 6 | 84 | |||
More Information
Set Membership: |
|

CHMP3
Genomic Map