Assay Details
| Gene Symbol: NAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045897.1 | NP_001039362.1 | 2 - 3 | 1111 | 55 | |
| GenBank mRNA | BC114714.1 | - | 2 - 3 | 1111 | 55 | |
More Information
Set Membership: |
|

NAB2
Genomic Map