Assay Details
| Gene Symbol: MRPL53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045926.2 | NP_001039391.1 | 2 - 3 | 222 | 99 | |
| GenBank mRNA | BC105487.1 | - | 2 - 3 | 223 | 99 | |
More Information
Set Membership: |
|

MRPL53
Genomic Map