Assay Details
| Gene Symbol: SIAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045968.2 | NP_001039433.1 | 9 - 10 | 1187 | 68 | |
| GenBank mRNA | BC112725.1 | - | 9 - 10 | 1188 | 68 | |
| BT030505.1 | - | 9 - 10 | 1210 | 68 | ||
More Information
Set Membership: |
|

SIAE
Genomic Map