Assay Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046427.2 | NP_001039892.1 | 7 - 8 | 560 | 94 | |
| GenBank mRNA | BC112679.1 | - | 7 - 8 | 561 | 94 | |
More Information
Set Membership: |
|

DNAL1
Genomic Map