Assay Details
| Gene Symbol: SLC7A6OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075567.1 | NP_001069035.1 | 4 - 5 | 800 | 65 | |
| GenBank mRNA | BC116007.1 | - | 4 - 5 | 800 | 65 | |
More Information
Set Membership: |
|

SLC7A6OS
Genomic Map