Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076026.2 | NP_001069494.1 | 1 - 2 | 240 | 54 | |
| GenBank mRNA | BC114775.1 | - | 1 - 2 | 242 | 54 | |
More Information
Set Membership: |
|

LIN37
Genomic Map