Assay Details
| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076137.1 | NP_001069605.1 | 2 - 3 | 442 | 57 | |
| GenBank mRNA | BC116085.1 | - | 2 - 3 | 442 | 57 | |
More Information
Set Membership: |
|

ANKRD54
Genomic Map