Assay Details
| Gene Symbol: CALB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076195.1 | NP_001069663.1 | 1 - 2 | 292 | 108 | |
| GenBank mRNA | BC116125.1 | - | 1 - 2 | 292 | 108 | |
More Information
Set Membership: |
|

CALB1
Genomic Map