Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076213.1 | NP_001069681.1 | 2 - 3 | 192 | 83 | |
| GenBank mRNA | BC118086.1 | - | 2 - 3 | 192 | 83 | |
| BT029824.1 | - | 2 - 3 | 193 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118086.1, NM_001076213.1 transcripts.
Set Membership: |
|

ZNF45
Genomic Map