Assay Details
| Gene Symbol: ZNF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076213.1 | NP_001069681.1 | 5 - 6 | 442 | 73 | |
| GenBank mRNA | BC118086.1 | - | 5 - 6 | 442 | 73 | |
| BT029824.1 | - | 5 - 6 | 443 | 73 | ||
More Information
Set Membership: |
|

ZNF45
Genomic Map