Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076222.2 | NP_001069690.1 | 2 - 3 | 207 | 62 | |
| GenBank mRNA | BC118414.1 | - | 2 - 3 | 208 | 62 | |
More Information
Set Membership: |
|

LMCD1
Genomic Map