Assay Details
| Gene Symbol: LHFPL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076226.1 | NP_001069694.1 | 1 - 2 | 130 | 141 | |
| GenBank mRNA | BC118076.1 | - | 1 - 2 | 130 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118076.1, NM_001076226.1 transcripts.
Set Membership: |
|

LHFPL4
Genomic Map