Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076291.1 | NP_001069759.1 | 6 - 7 | 703 | 71 | |
| GenBank mRNA | BC118262.1 | - | 6 - 7 | 703 | 71 | |
More Information
Set Membership: |
|

BAG1
Genomic Map