Assay Details
Gene Symbol: COA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076459.2 | NP_001069927.1 | 2 - 3 | 355 | 71 | |
GenBank mRNA | BC118424.1 | - | 2 - 3 | 357 | 71 | |
BT025435.1 | - | 2 - 3 | 329 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
