Assay Details
| Gene Symbol: SECTM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076529.1 | NP_001069997.1 | 6 - 7 | 937 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SECTM1A| Gene Symbol: SECTM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076529.1 | NP_001069997.1 | 6 - 7 | 937 | 82 | |
Set Membership: |
|