Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076977.2 | NP_001070445.1 | 2 - 3 | 86 | 99 | |
| GenBank mRNA | AM075813.1 | - | 2 - 3 | 87 | 99 | |
| BC142038.1 | - | 2 - 3 | 130 | 99 | ||
More Information
Set Membership: |
|

HCST
Genomic Map