Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076977.2 | NP_001070445.1 | 3 - 4 | 217 | 76 | |
| GenBank mRNA | AM075813.1 | - | 3 - 4 | 218 | 76 | |
| BC142038.1 | - | 3 - 4 | 261 | 76 | ||
More Information
Set Membership: |
|

HCST
Genomic Map