Assay Details
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174062.4 | NP_776487.1 | 1 | 485 | 61 | |
| GenBank mRNA | AB003093.1 | - | 2 - 3 | 425 | 61 | |
| AB098915.1 | - | 2 - 3 | 287 | 61 | ||
| AB098993.1 | - | 2 - 3 | 473 | 61 | ||
| AF540563.1 | - | 2 - 3 | 233 | 61 | ||
| AY911329.1 | - | 2 - 3 | 485 | 61 | ||
| BC105376.1 | - | 1 | 486 | 61 | ||
| BC151549.1 | - | 2 - 3 | 487 | 61 | ||
| DQ347567.1 | - | 1 - 2 | 142 | 61 | ||
| DQ347591.1 | - | 1 - 2 | 150 | 61 | ||
More Information
Set Membership: |
|

FTH1
Genomic Map