Assay Details
| Gene Symbol: PAICS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205813.2 | NP_991382.2 | 5 - 6 | 897 | 72 | |
| GenBank mRNA | AY344127.1 | - | 6 - 7 | 773 | 72 | |
| BC113344.1 | - | 5 - 6 | 897 | 72 | ||
| BT030591.2 | - | 5 - 6 | 739 | 72 | ||
More Information
Set Membership: |
|

PAICS
Genomic Map