Assay Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034380.2 | NP_001029552.2 | 1 - 2 | 237 | 65 | |
| GenBank mRNA | BT026308.1 | - | 1 - 2 | 237 | 65 | |
More Information
Set Membership: |
|

CFAP126
Genomic Map