Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075278.1 | NP_001068746.1 | 5 - 6 | 893 | 81 | |
| GenBank mRNA | BC123521.1 | - | 5 - 6 | 893 | 81 | |
More Information
Set Membership: |
|

ZDHHC3
Genomic Map