Assay Details
| Gene Symbol: METTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075288.2 | NP_001068756.2 | 9 - 10 | 1255 | 71 | |
| GenBank mRNA | BC120162.1 | - | 9 - 10 | 1257 | 71 | |
More Information
Set Membership: |
|

METTL8
Genomic Map