Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075314.1 | NP_001068782.1 | 3 - 4 | 283 | 56 | |
| GenBank mRNA | BC122844.1 | - | 3 - 4 | 283 | 56 | |
More Information
Set Membership: |
|

RNF126
Genomic Map