Assay Details
| Gene Symbol: CPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075372.2 | NP_001068840.1 | 7 - 8 | 832 | 82 | |
| GenBank mRNA | BC122774.1 | - | 7 - 8 | 833 | 82 | |
More Information
Set Membership: |
|

CPO
Genomic Map