Assay Details
| Gene Symbol: TERF2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075412.2 | NP_001068880.1 | 1 - 2 | 762 | 76 | |
| GenBank mRNA | BC120017.1 | - | 1 - 2 | 764 | 76 | |
More Information
Set Membership: |
|

TERF2IP
Genomic Map