Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075457.1 | NP_001068925.1 | 7 - 8 | 575 | 61 | |
| GenBank mRNA | BC118185.1 | - | 7 - 8 | 575 | 61 | |
More Information
Set Membership: |
|

CHMP6
Genomic Map