Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075492.1 | NP_001068960.1 | 11 - 12 | 1076 | 59 | |
| GenBank mRNA | BC119863.1 | - | 11 - 12 | 1076 | 59 | |
More Information
Set Membership: |
|

CCZ1
Genomic Map