Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075516.1 | NP_001068984.1 | 2 - 3 | 516 | 58 | |
| GenBank mRNA | BC120090.1 | - | 2 - 3 | 516 | 58 | |
More Information
Set Membership: |
|

CLDN18
Genomic Map