Assay Details
| Gene Symbol: MYLPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075647.1 | NP_001069115.1 | 5 - 6 | 400 | 67 | |
| GenBank mRNA | BC120431.1 | - | 5 - 6 | 400 | 67 | |
More Information
Set Membership: |
|

MYLPF
Genomic Map