Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075660.1 | NP_001069128.1 | 5 - 6 | 414 | 57 | |
| GenBank mRNA | BC120309.1 | - | 5 - 6 | 414 | 57 | |
More Information
Set Membership: |
|

DCTN3
Genomic Map