Assay Details
| Gene Symbol: WDR41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075690.1 | NP_001069158.1 | 10 - 11 | 1115 | 62 | |
| GenBank mRNA | BC120444.1 | - | 10 - 11 | 1115 | 62 | |
More Information
Set Membership: |
|

WDR41
Genomic Map