Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075726.1 | NP_001069194.1 | 2 - 3 | 482 | 62 | |
| GenBank mRNA | DQ832186.1 | - | 2 - 3 | 482 | 62 | |
More Information
Set Membership: |
|

HSD17B2
Genomic Map