Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075922.1 | NP_001069390.1 | 2 - 3 | 293 | 56 | |
| GenBank mRNA | BC123397.1 | - | 2 - 3 | 293 | 56 | |
More Information
Set Membership: |
|

DDIT4
Genomic Map