Assay Details
| Gene Symbol: CDPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076093.2 | NP_001069561.1 | 1 - 2 | 313 | 141 | |
| GenBank mRNA | BC120167.1 | - | 1 - 2 | 315 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120167.1, NM_001076093.2 transcripts.
Set Membership: |
|

CDPF1
Genomic Map