Assay Details
| Gene Symbol: CDPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076093.2 | NP_001069561.1 | 3 - 4 | 536 | 62 | |
| GenBank mRNA | BC120167.1 | - | 3 - 4 | 538 | 62 | |
More Information
Set Membership: |
|

CDPF1
Genomic Map