Assay Details
| Gene Symbol: F2RL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076138.2 | NP_001069606.1 | 1 - 2 | 136 | 143 | |
| GenBank mRNA | BC120099.1 | - | 1 - 2 | 137 | 143 | |
More Information
Set Membership: |
|

F2RL3
Genomic Map