Assay Details
| Gene Symbol: YIPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076219.1 | NP_001069687.1 | 1 - 2 | 193 | 91 | |
| GenBank mRNA | BC120404.1 | - | 1 - 2 | 193 | 91 | |
More Information
Set Membership: |
|

YIPF3
Genomic Map