Assay Details
| Gene Symbol: PSMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076347.1 | NP_001069815.1 | 5 - 6 | 685 | 68 | |
| GenBank mRNA | BC120126.1 | - | 5 - 6 | 685 | 68 | |
More Information
Set Membership: |
|

PSMG1
Genomic Map