Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076387.1 | NP_001069855.1 | 4 - 5 | 616 | 62 | |
| GenBank mRNA | BC120345.1 | - | 4 - 5 | 616 | 62 | |
More Information
Set Membership: |
|

TMEM38B
Genomic Map