Assay Details
| Gene Symbol: PGM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076903.1 | NP_001070371.1 | 10 - 11 | 1697 | 89 | |
| GenBank mRNA | BC123640.1 | - | 10 - 11 | 1697 | 89 | |
| BC149239.1 | - | 10 - 11 | 1968 | 89 | ||
More Information
Set Membership: |
|

PGM1
Genomic Map