Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076914.1 | NP_001070382.2 | 4 - 5 | 378 | 133 | |
| GenBank mRNA | BC123537.1 | - | 4 - 5 | 378 | 133 | |
More Information
Set Membership: |
|

ZNF527
Genomic Map