Assay Details
| Gene Symbol: THSD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078030.3 | NP_001071498.2 | 3 - 4 | 665 | 75 | |
| GenBank mRNA | BC120293.1 | - | 3 - 4 | 568 | 75 | |
More Information
Set Membership: |
|

THSD4
Genomic Map