Assay Details
| Gene Symbol: ZYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078101.2 | NP_001071569.1 | 2 - 3 | 282 | 60 | |
| GenBank mRNA | BC123619.1 | - | 2 - 3 | 283 | 60 | |
More Information
Set Membership: |
|

ZYX
Genomic Map