Assay Details
| Gene Symbol: GNRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078137.1 | NP_001071605.1 | 2 - 3 | 207 | 75 | |
| GenBank mRNA | BC120472.1 | - | 2 - 3 | 207 | 75 | |
More Information
Set Membership: |
|

GNRH1
Genomic Map