Assay Details
| Gene Symbol: OPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079605.1 | NP_001073073.1 | 1 - 2 | 172 | 93 | |
| GenBank mRNA | BC122701.1 | - | 1 - 2 | 172 | 93 | |
More Information
Set Membership: |
|

OPA3
Genomic Map