Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080235.2 | NP_001073704.1 | 6 - 7 | 920 | 64 | |
| GenBank mRNA | BC122830.1 | - | 6 - 7 | 921 | 64 | |
More Information
Set Membership: |
|

TIMMDC1
Genomic Map