Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080365.2 | NP_001073834.1 | 1 - 2 | 170 | 133 | |
| GenBank mRNA | BC120279.1 | - | 1 - 2 | 171 | 133 | |
More Information
Set Membership: |
|

ARRDC1
Genomic Map